2-Fluor-3-methoxybenzoesäure 99%
Lieferant: Apollo Scientific
|
Achtung
|
PC9295-25G
PC9295-5G
PC9295-100G
APOSPC9295-25GEA
137
EUR
APOSPC9295-25G
APOSPC9295-5G
APOSPC9295-100G
2-Fluor-3-methoxybenzoesäure 99%
2-Fluoro-3-methoxybenzoic acid
Formel:
C₈H₇FO₃ Schmelzpunkt: 157…161 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02179621 CAS-Nummer: 137654-20-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...