4-Brom-3-(trifluormethyl)benzoesäure

Lieferant: Apollo Scientific
Achtung

PC9303-1G PC9303-25G PC9303-5G
APOSPC9303-1GEA 21.9 EUR
APOSPC9303-1G APOSPC9303-25G APOSPC9303-5G
4-Brom-3-(trifluormethyl)benzoesäure
4-Bromo-3-(trifluoromethyl)benzoic acid
Formel: C₈H₄BrF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08459288
CAS-Nummer: 161622-14-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR