Ethyl-2-(3-{[3-(trifluoromethyl)anilino]carbonyl}-2-pyridinyl)acetate 97%

Lieferant: Apollo Scientific
Achtung

PC9315-1G PC9315-250MG
APOSPC9315-1GEA 258 EUR
APOSPC9315-1G APOSPC9315-250MG
Ethyl-2-(3-{[3-(trifluoromethyl)anilino]carbonyl}-2-pyridinyl)acetate 97%
Ethyl-2-(3-{[3-(trifluoromethyl)anilino]carbonyl}-2-pyridinyl)acetate
Formel: C₁₇H₁₅F₃N₂O₃
Molecular Weight: 352,32 g/mol
Schmelzpunkt: 106…108 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00974992
CAS-Nummer: 338392-12-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR