1-(4-Fluorobenzyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile 97%

Lieferant: Apollo Scientific

PC9332-1G PC9332-500MG PC9332-5G
APOSPC9332-1GEA 275 EUR
APOSPC9332-1G APOSPC9332-500MG APOSPC9332-5G
1-(4-Fluorobenzyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile 97%
1-(4-Fluorobenzyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile
Formel: C₁₄H₁₃FN₂O
Molecular Weight: 244.27 g/mol
Schmelzpunkt: 108…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00975039
CAS-Nummer: 338748-69-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR