3-[(4-Fluorobenzyl)oxy]-2-thiophenecarbohydrazide 97%

Lieferant: Apollo Scientific

PC9334-1G PC9334-500MG PC9334-5G
APOSPC9334-1GEA 275 EUR
APOSPC9334-1G APOSPC9334-500MG APOSPC9334-5G
3-[(4-Fluorobenzyl)oxy]-2-thiophenecarbohydrazide 97%
3-[(4-Fluorobenzyl)oxy]-2-thiophenecarbohydrazide
Formel: C₁₂H₁₁FN₂O₂S
Molecular Weight: 266,3 g/mol
Schmelzpunkt: 107…109 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00215161
CAS-Nummer: 338751-50-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR