{2-[(4-Fluorobenzyl)sulphanyl]-1-methyl-1H-imidazol-5-yl}methanol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(4-Fluorobenzythio)-5-(hydroxymethyl)-1-methyl-1H-imidazole

PC9337-1G PC9337-250MG
APOSPC9337-1GEA 441 EUR
APOSPC9337-1G APOSPC9337-250MG
{2-[(4-Fluorobenzyl)sulphanyl]-1-methyl-1H-imidazol-5-yl}methanol 97%
{2-[(4-Fluorobenzyl)sulphanyl]-1-methyl-1H-imidazol-5-yl}methanol
Formel: C₁₂H₁₃FN₂OS
Molecular Weight: 252.31 g/mol
Schmelzpunkt: 124…126 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00215148
CAS-Nummer: 338422-40-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR