3-Chlor-4-fluorphenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Dihydroxy(3-chlor-4-fluorphenyl)boran, (3-Chlor-4-fluorphenyl)metaborsäure, 3-Chlor-4-fluorbenzolboronsäure
PC9345-5G
PC9345-1G
PC9345-10G
APOSPC9345-5GEA
21.9
EUR
APOSPC9345-5G
APOSPC9345-1G
APOSPC9345-10G
3-Chlor-4-fluorphenylboronsäure
3-Chlor-4-fluorphenylboronsäure
Formel:
C₆H₅BClFO₂ Molecular Weight: 174,37 g/mol Schmelzpunkt: 255…260 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00051800 CAS-Nummer: 144432-85-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...