1-(4-Fluorophenyl)-2-{[5-(hydroxymethyl)-1-methyl-1H-imidazol-2-yl]sulphanyl}-1-ethanone

Lieferant: Apollo Scientific

Synonyme: 4'-Fluoro-2-[5-(hydroxymethyl)-1-methyl-1H-imidazol-2-ylthio]acetophenone

PC9357-1G PC9357-500MG PC9357-5G
APOSPC9357-1GEA 533 EUR
APOSPC9357-1G APOSPC9357-500MG APOSPC9357-5G
1-(4-Fluorophenyl)-2-{[5-(hydroxymethyl)-1-methyl-1H-imidazol-2-yl]sulphanyl}-1-ethanone
1-(4-Fluorophenyl)-2-{[5-(hydroxymethyl)-1-methyl-1H-imidazol-2-yl]sulphanyl}-1-ethanone
Formel: C₁₃H₁₃FN₂O₂S
Molecular Weight: 280,32 g/mol
Schmelzpunkt: 121…123 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00215160
CAS-Nummer: 338422-62-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR