4-[(4-Fluorophenyl)sulphanyl]aniline 97%

Lieferant: Apollo Scientific

Synonyme: 4-(4-Fluorophenylthio)aniline

PC9366-1G PC9366-500MG PC9366-5G
APOSPC9366-1GEA 270 EUR
APOSPC9366-1G APOSPC9366-500MG APOSPC9366-5G
4-[(4-Fluorophenyl)sulphanyl]aniline 97%
4-[(4-Fluorophenyl)sulphanyl]aniline
Formel: C₁₂H₁₀FNS
Schmelzpunkt: 60…62 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00172068
CAS-Nummer: 24900-69-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR