2,4,5-Trifluorzimtsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,4,5-Trifluorcinnamsäure, 3-(2,4,5-Trifluorphenyl)propensäure
PC9367-1G
PC9367-10G
PC9367-5G
APOSPC9367-1GEA
21.9
EUR
APOSPC9367-1G
APOSPC9367-10G
APOSPC9367-5G
2,4,5-Trifluorzimtsäure 98%
2,4,5-Trifluorzimtsäure
Formel:
C₉H₅F₃O₂ Molecular Weight: 202.13 g/mol Schmelzpunkt: 184…188 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061211 CAS-Nummer: 247170-17-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...