2-{[(4-Fluorophenyl)sulphonyl]amino}-4-methylpentanoic acid 97%

Lieferant: Apollo Scientific

Synonyme: 2-(4-Fluorobenzenesulphamido)-4-methylpentanoic acid

PC9373-1G PC9373-500MG PC9373-5G
APOSPC9373-1GEA 217 EUR
APOSPC9373-1G APOSPC9373-500MG APOSPC9373-5G
2-{[(4-Fluorophenyl)sulphonyl]amino}-4-methylpentanoic acid 97%
2-{[(4-Fluorophenyl)sulphonyl]amino}-4-methylpentanoic acid
Formel: C₁₂H₁₆FNO₄S
Molecular Weight: 289,33 g/mol
Schmelzpunkt: 124…126 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00172812
CAS-Nummer: 251097-25-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR