1-(4-Fluorphenylsulfonyl)pyrrolidin-2-carbonsäure

Lieferant: Apollo Scientific
Achtung

PC9380-1G PC9380-5G PC9380-250MG
APOSPC9380-1GEA 49.3 EUR
APOSPC9380-1G APOSPC9380-5G APOSPC9380-250MG
1-(4-Fluorphenylsulfonyl)pyrrolidin-2-carbonsäure
1-(4-Fluorphenylsulfonyl)pyrrolidin-2-carbonsäure
Formel: C₁₁H₁₂FNO₄S
Schmelzpunkt: 96…99 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00172808
CAS-Nummer: 251096-96-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR