2-({Imino[3-(trifluoromethyl)phenyl]methyl}amino)acetic acid 97%
Lieferant: Apollo Scientific
Synonyme:
N-(2-Carboxymethyl)-3-(trifluoromethyl)benzamidine
PC9385-1G
PC9385-500MG
PC9385-5G
APOSPC9385-1GEA
275
EUR
APOSPC9385-1G
APOSPC9385-500MG
APOSPC9385-5G
2-({Imino[3-(trifluoromethyl)phenyl]methyl}amino)acetic acid 97%
2-({Imino[3-(trifluoromethyl)phenyl]methyl}amino)acetic acid
Formel:
C₁₀H₉F₃N₂O₂ Molecular Weight: 246,19 g/mol Schmelzpunkt: 204…208 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01935986 CAS-Nummer: 338982-13-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...