Methyl-4-[2-amino-4-(trifluoromethyl)anilino]butanoate 97%

Lieferant: Apollo Scientific

PC9391-1G PC9391-500MG PC9391-5G
APOSPC9391-1GEA 533 EUR
APOSPC9391-1G APOSPC9391-500MG APOSPC9391-5G
Methyl-4-[2-amino-4-(trifluoromethyl)anilino]butanoate 97%
Methyl-4-[2-amino-4-(trifluoromethyl)anilino]butanoate
Formel: C₁₂H₁₅F₃N₂O₂
Molecular Weight: 276.26 g/mol
Schmelzpunkt: 91…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00793102
CAS-Nummer: 339101-34-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR