Methyl-3-(2-chloro-6-fluorophenyl)-5-[2-(dimethylamino)vinyl]-4-isoxazolecarboxylate

Lieferant: Apollo Scientific
Achtung

PC9395-1G PC9395-250MG
APOSPC9395-1GEA 186 EUR
APOSPC9395-1G APOSPC9395-250MG
Methyl-3-(2-chloro-6-fluorophenyl)-5-[2-(dimethylamino)vinyl]-4-isoxazolecarboxylate
Methyl-3-(2-chloro-6-fluorophenyl)-5-[2-(dimethylamino)vinyl]-4-isoxazolecarboxylate
Formel: C₁₅H₁₄ClFN₂O₃
Molecular Weight: 324.74 g/mol
Schmelzpunkt: 139…141 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00127467
CAS-Nummer: 338417-50-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR