Methyl-4-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]amino}butanoate
Lieferant: Apollo Scientific
|
Achtung
|
PC9400-1G
PC9400-250MG
APOSPC9400-1GEA
325
EUR
APOSPC9400-1G
APOSPC9400-250MG
Methyl-4-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]amino}butanoate
Methyl-4-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]amino}butanoate
Formel:
C₁₁H₁₂ClF₃N₂O₂ Molecular Weight: 296,68 g/mol Schmelzpunkt: 46…48 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00172609 CAS-Nummer: 332361-10-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...