3-(Chlormethyl)-5-(3-(trifluormethyl)phenyl)-1,2,4-oxadiazol 95+%
Lieferant: Apollo Scientific
|
Gefahr
|
PC9405-1G
PC9405-5G
APOSPC9405-1GEA
174
EUR
APOSPC9405-1G
APOSPC9405-5G
3-(Chlormethyl)-5-(3-(trifluormethyl)phenyl)-1,2,4-oxadiazol 95+%
3-(Chloromethyl)-5-(3-(trifluoromethyl)phenyl)-1,2,4-oxadiazole
Formel:
C₁₀H₆ClF₃N₂O Schmelzpunkt: 34…35 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00084969 CAS-Nummer: 175205-63-7 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...