Methyl-1-(4-fluorobenzoyl)-4-hydroxy-2-pyrrolidinecarboxylate 97%

Lieferant: Apollo Scientific

PC9408-1G PC9408-250MG PC9408-500MG
APOSPC9408-1GEA 538 EUR
APOSPC9408-1G APOSPC9408-250MG APOSPC9408-500MG
Methyl-1-(4-fluorobenzoyl)-4-hydroxy-2-pyrrolidinecarboxylate 97%
Methyl-1-(4-fluorobenzoyl)-4-hydroxy-2-pyrrolidinecarboxylate
Formel: C₁₃H₁₄FNO₄
Molecular Weight: 267,26 g/mol
Schmelzpunkt: 150…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00202776
CAS-Nummer: 318247-24-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR