2-Amino-4,6-difluorphenol 95+%
Lieferant: Apollo Scientific
|
Achtung
|
PC9412-1G
PC9412-250MG
APOSPC9412-1GEA
227
EUR
APOSPC9412-1G
APOSPC9412-250MG
2-Amino-4,6-difluorphenol 95+%
2-Amino-4,6-difluorphenol
Formel:
C₆H₅F₂NO Molecular Weight: 145,1068 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01862077 CAS-Nummer: 133788-83-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...