Methyl-4-(4-fluorophenoxy)-3-nitrobenzenecarboxylate 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 4-(4-fluorophenoxy)-3-nitrobenzoate
PC9414-1G
PC9414-250MG
APOSPC9414-1GEA
383
EUR
APOSPC9414-1G
APOSPC9414-250MG
Methyl-4-(4-fluorophenoxy)-3-nitrobenzenecarboxylate 97%
Methyl-4-(4-fluorophenoxy)-3-nitrobenzenecarboxylate
Formel:
C₁₄H₁₀FNO₅ Molecular Weight: 291,24 g/mol Schmelzpunkt: 101…103 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00138765 CAS-Nummer: 320417-11-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...