1-(2-Cyanoethyl)-3-(trifluoromethyl)pyrid-2-(1H)-one 97%

Lieferant: Apollo Scientific

PC9420-1G PC9420-10G
APOSPC9420-1GEA 87.3 EUR
APOSPC9420-1G APOSPC9420-10G
1-(2-Cyanoethyl)-3-(trifluoromethyl)pyrid-2-(1H)-one 97%
1-(2-Cyanoethyl)-3-(trifluoromethyl)pyrid-2-(1H)-one
Formel: C₉H₇F₃N₂O
Molecular Weight: 216,16 g/mol
Schmelzpunkt: 88…89 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00204216
CAS-Nummer: 175277-60-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR