2-{4-[2-Nitro-4-(trifluoromethyl)phenyl]piperazino}-1-ethanol 97%

Lieferant: Apollo Scientific

PC9468-1G PC9468-500MG PC9468-5G
APOSPC9468-1GEA 172 EUR
APOSPC9468-1G APOSPC9468-500MG APOSPC9468-5G
2-{4-[2-Nitro-4-(trifluoromethyl)phenyl]piperazino}-1-ethanol 97%
2-{4-[2-Nitro-4-(trifluoromethyl)phenyl]piperazino}-1-ethanol
Formel: C₁₃H₁₆F₃N₃O₃
Molecular Weight: 319.29 g/mol
Schmelzpunkt: 108…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01173874
CAS-Nummer: 330633-81-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR