4-Chlor-2-fluorphenylacetonitril 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Chloro-2-fluorobenzeneacetonitrile
PC9472-25G
PC9472-5G
PC9472-100G
APOSPC9472-25GEA
55.1
EUR
APOSPC9472-25G
APOSPC9472-5G
APOSPC9472-100G
4-Chlor-2-fluorphenylacetonitril 98%
4-Chlor-2-fluorphenylacetonitril
Formel:
C₈H₅ClFN Molecular Weight: 169,59 g/mol Schmelzpunkt: 36…39 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01312758 CAS-Nummer: 75279-53-7 UN: 3439 ADR: 6.1,III |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...