2-Phenyl-2-({[3-(trifluoromethyl)phenyl]sulphonyl}amino)acetic acid 97%

Lieferant: Apollo Scientific

Synonyme: N-{[3-(Trifluoromethyl)phenyl]sulphonyl}-DL-phenylglycine

PC9482-1G PC9482-500MG PC9482-5G
APOSPC9482-1GEA 275 EUR
APOSPC9482-1G APOSPC9482-500MG APOSPC9482-5G
2-Phenyl-2-({[3-(trifluoromethyl)phenyl]sulphonyl}amino)acetic acid 97%
2-Phenyl-2-({[3-(trifluoromethyl)phenyl]sulphonyl}amino)acetic acid
Formel: C₁₅H₁₂F₃NO₄S
Schmelzpunkt: 169…171 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00172802
CAS-Nummer: 250714-41-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR