4-Brom-2,5-difluoranilin 98%

Lieferant: Apollo Scientific
Achtung

PC9483-100G PC9483-25G PC9483-5G
APOSPC9483-100GEA 188 EUR
APOSPC9483-100G APOSPC9483-25G APOSPC9483-5G
4-Brom-2,5-difluoranilin 98%
4-Brom-2,5-difluoranilin
Formel: C₆H₄BrF₂N
Molecular Weight: 208,01 g/mol
Schmelzpunkt: 74…75 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01861125
CAS-Nummer: 112279-60-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR