3-{[3-(Trifluoromethyl)benzyl]oxy}-2-thiophenecarboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

PC9490-1G PC9490-500MG
APOSPC9490-1GEA 467 EUR
APOSPC9490-1G APOSPC9490-500MG
3-{[3-(Trifluoromethyl)benzyl]oxy}-2-thiophenecarboxylic acid 97%
3-{[3-(Trifluoromethyl)benzyl]oxy}-2-thiophenecarboxylic acid
Formel: C₁₃H₉F₃O₃S
Molecular Weight: 302,27 g/mol
Schmelzpunkt: 118…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173491
CAS-Nummer: 339009-59-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR