4-({[3-(Trifluoromethyl)phenyl]sulphanyl}methyl)aniline 97%
Lieferant: Apollo Scientific
Synonyme:
3-(4-Aminobenzylthio)benzotrifluoride
PC9499-1G
PC9499-500MG
APOSPC9499-1GEA
666
EUR
APOSPC9499-1G
APOSPC9499-500MG
4-({[3-(Trifluoromethyl)phenyl]sulphanyl}methyl)aniline 97%
4-({[3-(Trifluoromethyl)phenyl]sulphanyl}methyl)aniline
Formel:
C₁₄H₁₂F₃NS Molecular Weight: 283,32 g/mol Schmelzpunkt: 68…71 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173208 CAS-Nummer: 338398-58-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...