2-Fluor-5-hydroxybenzoesäure
Lieferant: Apollo Scientific
|
Achtung
|
PC9534-1G
PC9534-5G
PC9534-10G
APOSPC9534-1GEA
21.9
EUR
APOSPC9534-1G
APOSPC9534-5G
APOSPC9534-10G
2-Fluor-5-hydroxybenzoesäure
2-Fluoro-5-hydroxybenzoic acid
Formel:
C₇H₅FO₃ Schmelzpunkt: 204…206 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01862084 CAS-Nummer: 51446-30-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...