3-Methoxy-5-(trifluormethyl)benzonitril 97%

Lieferant: Apollo Scientific
Achtung

PC9552-1G PC9552-25G PC9552-5G
APOSPC9552-1GEA 21.9 EUR
APOSPC9552-1G APOSPC9552-25G APOSPC9552-5G
3-Methoxy-5-(trifluormethyl)benzonitril 97%
3-Methoxy-5-(trifluormethyl)benzonitril
Formel: C₉H₆F₃NO
Molecular Weight: 201,1453 g/mol
Schmelzpunkt: 38 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08458145
CAS-Nummer: 868167-61-7
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR