5-Fluorobenzoxazole 97+%

Lieferant: Apollo Scientific
Achtung

PC9577-1G PC9577-250MG PC9577-5G
APOSPC9577-1GEA 78.3 EUR
APOSPC9577-1G APOSPC9577-250MG APOSPC9577-5G
5-Fluorobenzoxazole 97+%
5-Fluorobenzoxazole
Formel: C₇H₄FNO
Molecular Weight: 137,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08458151
CAS-Nummer: 221347-71-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR