1-(3-Chlor-4-fluorphenyl)biguanid Hydrochlorid

Lieferant: Apollo Scientific
Achtung

PC9620-1G PC9620-25G PC9620-5G
APOSPC9620-1GEA 42.2 EUR
APOSPC9620-1G APOSPC9620-25G APOSPC9620-5G
1-(3-Chlor-4-fluorphenyl)biguanid Hydrochlorid
1-(3-Chlor-4-fluorphenyl)biguanid Hydrochlorid
Formel: C₈H₁₀Cl₂FN₅
Schmelzpunkt: 209…213 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00126332
CAS-Nummer: 915071-41-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR