2-Chlor-3-fluorbenzonitril 97%

Lieferant: Apollo Scientific
Achtung

PC9671-5G PC9671-25G
APOSPC9671-5GEA 33.5 EUR
APOSPC9671-5G APOSPC9671-25G
2-Chlor-3-fluorbenzonitril 97%
2-Chlor-3-fluorbenzonitril
Formel: C₇H₃ClFN
Molecular Weight: 155,56 g/mol
Schmelzpunkt: 55…56 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03407966
CAS-Nummer: 874781-08-5
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR