6-Fluoroindole-3-carboxaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Fluoro-1H-indole-3-carbaldehyde

PC9685-25G PC9685-5G PC9685-100G
APOSPC9685-25GEA 184 EUR
APOSPC9685-25G APOSPC9685-5G APOSPC9685-100G
6-Fluoroindole-3-carboxaldehyde
6-Fluoroindole-3-carboxaldehyde
Formel: C₉H₆FNO
Molecular Weight: 163.15 g/mol
Schmelzpunkt: 173…176 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00069703
CAS-Nummer: 2795-41-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR