1-(4-Fluorphenyl)-2-pyrrolidinon 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(4-Fluorophenyl)pyrrolidin-2-one
PC9696-1G
PC9696-5G
PC9696-250MG
APOSPC9696-1GEA
94.5
EUR
APOSPC9696-1G
APOSPC9696-5G
APOSPC9696-250MG
1-(4-Fluorphenyl)-2-pyrrolidinon 95+%
1-(4-Fluorphenyl)-2-pyrrolidinon
Formel:
C₁₀H₁₀FNO Molecular Weight: 179,19 g/mol Schmelzpunkt: 54…57 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00138504 CAS-Nummer: 54660-08-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...