4-Amino-2,6-difluoropyridine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,6-Difluoropyridin-4-amine

PC9706-1G PC9706-5G PC9706-250MG
APOSPC9706-1GEA 82.8 EUR
APOSPC9706-1G APOSPC9706-5G APOSPC9706-250MG
4-Amino-2,6-difluoropyridine 97%
4-Amino-2,6-difluoropyridine
Formel: C₅H₄F₂N₂
Molecular Weight: 130.098 g/mol
Schmelzpunkt: 124…128 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08436089
CAS-Nummer: 63489-58-7
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR