4-Bromo-2-chloro-5-fluorobenzoic acid

Lieferant: Apollo Scientific
Achtung

PC9723-1G PC9723-25G PC9723-5G
APOSPC9723-1GEA 21.9 EUR
APOSPC9723-1G APOSPC9723-25G APOSPC9723-5G
4-Bromo-2-chloro-5-fluorobenzoic acid
4-Bromo-2-chloro-5-fluorobenzoic acid
Formel: C₇H₃BrClFO₂
Molecular Weight: 253.451 g/mol
Schmelzpunkt: 150…155 °C
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00236589
CAS-Nummer: 177480-81-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR