4-(4-Fluorphenoxy)anilin

Lieferant: Apollo Scientific
Gefahr

PC9750-1G PC9750-25G PC9750-5G
APOSPC9750-1GEA 21.9 EUR
APOSPC9750-1G APOSPC9750-25G APOSPC9750-5G
4-(4-Fluorphenoxy)anilin
4-(4-Fluorphenoxy)anilin
Formel: C₁₂H₁₀FNO
Schmelzpunkt: 60…64 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01312350
CAS-Nummer: 36160-82-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR