4-Amino-3-fluorbenzoesäure 97+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Amino-3-fluorobenzoic Acid

PC9752-1G PC9752-5G
APOSPC9752-1GEA 21.9 EUR
APOSPC9752-1G APOSPC9752-5G
4-Amino-3-fluorbenzoesäure 97+%
4-Amino-3-fluorobenzoic acid
Formel: C₇H₆FNO₂
Molecular Weight: 155.13 g/mol
Schmelzpunkt: 212…218 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01660374
CAS-Nummer: 455-87-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR