3-Fluorbenzophenon 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: m-Fluorbenzophenon

PC9753-25G PC9753-5G
APOSPC9753-25GEA 252 EUR
APOSPC9753-25G APOSPC9753-5G
3-Fluorbenzophenon 98%
3-Fluorbenzophenon
Formel: C₁₃H₉FO
Molecular Weight: 200,21 g/mol
Schmelzpunkt: 48…52 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00083553
CAS-Nummer: 345-69-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR