3-Bromo-1,1,1-trifluoroacetone oxime

Lieferant: Apollo Scientific
Achtung

PC9757-1G PC9757-5G
APOSPC9757-1GEA 136 EUR
APOSPC9757-1G APOSPC9757-5G
3-Bromo-1,1,1-trifluoroacetone oxime
3-Bromo-1,1,1-trifluoroacetone oxime
Formel: C₃H₃BrF₃NO
Molecular Weight: 205.97 g/mol
Siedepunkt: 80…90 °C/85mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD04038272
CAS-Nummer: 117341-57-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR