2-Quinolin-2-yl-1-[3-(trifluoromethyl)phenyl]ethanone 95+%

Lieferant: Apollo Scientific

PC9770-25G PC9770-5G PC9770-1G
APOSPC9770-25GEA 1260 EUR
APOSPC9770-25G APOSPC9770-5G APOSPC9770-1G
2-Quinolin-2-yl-1-[3-(trifluoromethyl)phenyl]ethanone 95+%
2-Quinolin-2-yl-1-[3-(trifluoromethyl)phenyl]ethanone
Formel: C₁₈H₁₂F₃NO
Molecular Weight: 315.3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04037917
CAS-Nummer: 849021-38-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR