2-Cyan-6-fluoranilin 97%

Lieferant: Apollo Scientific
Achtung

PC9826-1G PC9826-25G PC9826-5G
APOSPC9826-1GEA 21.9 EUR
APOSPC9826-1G APOSPC9826-25G APOSPC9826-5G
2-Cyan-6-fluoranilin 97%
2-Cyan-6-fluoranilin
Formel: C₇H₅FN₂
Molecular Weight: 136,13 g/mol
Schmelzpunkt: 79…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368774
CAS-Nummer: 115661-37-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR