3-(Trifluormethyl)anthranilsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-3-(trifluormethyl)benzoesäure, 2-Amino-α,α,α-trifluor-m-toluylsäure

PC9853-25G PC9853-5G PC9853-1G
APOSPC9853-25GEA 145 EUR
APOSPC9853-25G APOSPC9853-5G APOSPC9853-1G
3-(Trifluormethyl)anthranilsäure 98%
3-(Trifluormethyl)anthranilsäure

Starting material for preparation of benzoisothiazolone & dithiobis(benzamide) antimicrobials.

Formel: C₈H₆F₃NO₂
Molecular Weight: 205.14 g/mol
Schmelzpunkt: 157…160 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00274210
CAS-Nummer: 313-12-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR