3-Fluor-4-hydroxybenzonitril

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Fluor-4-hydroxybenzoesäurenitril

PC9903-25G PC9903-5G
APOSPC9903-25GEA 46.4 EUR
APOSPC9903-25G APOSPC9903-5G
3-Fluor-4-hydroxybenzonitril
3-Fluor-4-hydroxybenzonitril
Formel: C₇H₄FNO
Molecular Weight: 137,11 g/mol
Schmelzpunkt: 132…133 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00215834
CAS-Nummer: 405-04-9
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR