4-Cyan-2-fluorbenzaldehyd 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Fluoro-4-formylbenzonitrile

PC9916-5G PC9916-1G PC9916-25G
APOSPC9916-5GEA 55.1 EUR
APOSPC9916-5G APOSPC9916-1G APOSPC9916-25G
4-Cyan-2-fluorbenzaldehyd 98%
4-Cyano-2-fluorobenzaldehyde
Formel: C₈H₄FNO
Molecular Weight: 149,1200 g/mol
Schmelzpunkt: 78…79 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368786
CAS-Nummer: 105942-10-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR