1,2-Dibrom-3-fluorbenzol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,3-Dibromofluorobenzene
PC9919-1G
PC9919-25G
PC9919-5G
APOSPC9919-1GEA
29
EUR
APOSPC9919-1G
APOSPC9919-25G
APOSPC9919-5G
1,2-Dibrom-3-fluorbenzol 98%
1,2-Dibromo-3-fluorobenzene
Formel:
C₆H₃Br₂F Molecular Weight: 253,8944 g/mol Schmelzpunkt: 15…20 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368789 CAS-Nummer: 811711-33-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...