1-(4-Nitro-2-(trifluoromethyl)phenyl)piperazine
Lieferant: Apollo Scientific
PC9931-10G
PC9931-1G
PC9931-5G
APOSPC9931-10GEA
241
EUR
APOSPC9931-10G
APOSPC9931-1G
APOSPC9931-5G
1-(4-Nitro-2-(trifluoromethyl)phenyl)piperazine
1-(4-Nitro-2-(trifluoromethyl)phenyl)piperazine
Formel:
C₁₁H₁₂F₃N₃O₂ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
381242-61-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...