Ethyl-2-(benzoyl-3-chloro-4-fluoroanilino)propanoate

Lieferant: Apollo Scientific

PC9936-1G PC9936-500MG PC9936-5G
APOSPC9936-1GEA 217 EUR
APOSPC9936-1G APOSPC9936-500MG APOSPC9936-5G
Ethyl-2-(benzoyl-3-chloro-4-fluoroanilino)propanoate
Ethyl-2-(benzoyl-3-chloro-4-fluoroanilino)propanoate
Formel: C₁₈H₁₇ClFNO₃
Molecular Weight: 349.79 g/mol
Schmelzpunkt: 86…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01310648
CAS-Nummer: 34029-27-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR