1-Brom-2-fluor-3-nitrobenzol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Brom-2-fluornitrobenzol

PC9945-5G PC9945-100G PC9945-25G
APOSPC9945-5GEA 30.6 EUR
APOSPC9945-5G APOSPC9945-100G APOSPC9945-25G
1-Brom-2-fluor-3-nitrobenzol 98%
1-Bromo-2-fluoro-3-nitrobenzene
Formel: C₆H₃BrFNO₂
Molecular Weight: 220 g/mol
Schmelzpunkt: 29…31 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05663754
CAS-Nummer: 58534-94-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR