2-Fluor-4-hydroxybenzoesäure

Lieferant: Apollo Scientific
Achtung

PC9958-100G PC9958-25G
APOSPC9958-100GEA 260 EUR
APOSPC9958-100G APOSPC9958-25G
2-Fluor-4-hydroxybenzoesäure
2-Fluoro-4-hydroxybenzoic acid
Formel: C₇H₅FO₃
Molecular Weight: 156,1112 g/mol
Schmelzpunkt: 199…202 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01310985
CAS-Nummer: 65145-13-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR